Please login to your online account to display your discounted pricing

R&D Systems Human UbcH5a/UBE2D1 Recombinant Protein


Manufacturer: r&d systems  RDSE2616050

 View more versions of this product

Catalog No. E2616050

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, Glycerol.
Gene ID (Entrez) 7321
Molecular Weight 17 kDa
Name UbcH5a/UBE2D1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 17 kDa
Formulation: Supplied as a solution in HEPES, NaCl, Glycerol.
Entrez Gene ID: 7321
Regulatory Status: RUO