Please login to your online account to display your discounted pricing

R&D Systems Human UbcH2/UBE2H Recombinant Protein


Manufacturer: r&d systems  RDSE2607100

 View more versions of this product

Catalog No. E2607100

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7328
Name UbcH2/UBE2H
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Molecular Weight 22 kDa
Source Human
Quantity 100μg
Conjugate Unlabeled
Purity 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes