Please login to your online account to display your discounted pricing

R&D Systems Human UbcH2/UBE2H Recombinant Protein

$175.98 - $293.04

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7328
Molecular Weight 22 kDa
Name UbcH2/UBE2H
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2607100

 
r&d systems
RDSE2607100
100μg Each for $293.04

E2607050

 
r&d systems
RDSE2607050
50μg Each for $175.98
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7328
Molecular Weight 22 kDa
Name UbcH2/UBE2H
Purity 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 22 kDa
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7328
Regulatory Status: RUO