missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TrkC Fc Chimera Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4916
Molecular Weight 72 kDa (monomer)
Name TrkC Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
373TC050 R&D Systems
RDS373TC050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
373TC050CF R&D Systems
RDS373TC050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Cys32-Asp428
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 72 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4916
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
72 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
4916
TrkC Fc Chimera
50μg
Human