Please login to your online account to display your discounted pricing

R&D Systems Human TRAIL/TNFSF10 Recombinant Protein, (E. coli-expressed)

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8743
Molecular Weight 19 kDa (monomer)
Name TRAIL/TNFSF10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

375TEC010CF

 
r&d systems
RDS375TEC010CF
Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4. Each for $415.83

375TEC010

 
r&d systems
RDS375TEC010
Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4 with BSA as a carrier protein. Each for $415.83
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8743
Molecular Weight 19 kDa (monomer)
Name TRAIL/TNFSF10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: E. coli-derived, Val114-Gly281, with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4.
Entrez Gene ID: 8743
Regulatory Status: RUO