Please login to your online account to display your discounted pricing

R&D Systems Human TRAIL/TNFSF10 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8743
Molecular Weight 21 kDa (monomer)
Name TRAIL/TNFSF10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

375TL010CF

 
r&d systems
RDS375TL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $415.83

375TL010

 
r&d systems
RDS375TL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $415.83
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8743
Molecular Weight 21 kDa (monomer)
Name TRAIL/TNFSF10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Thr95-Gly281, with an N-terminal Met and 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8743
Regulatory Status: RUO