missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TRAIL R3/TNFRSF10C Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8794
Molecular Weight 48 kDa (monomer)
Name TRAIL R3/TNFRSF10C Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
630TR100 R&D Systems
RDS630TR100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94
630TR100CF R&D Systems
RDS630TR100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala26-Ala221
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 48 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8794
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
48 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8794
TRAIL R3/TNFRSF10C Fc Chimera
100μg
Human