missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TRAIL R1/TNFRSF10A Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8797
Molecular Weight 43 kDa (monomer)
Name TRAIL R1/TNFRSF10A Fc Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
347DR100CF R&D Systems
RDS347DR100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
347DR100 R&D Systems
RDS347DR100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala109-Asn239 & Thr110-Asn239
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 43 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8797
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
43 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8797
TRAIL R1/TNFRSF10A Fc Chimera
100μg
Human