missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TNF RI/TNFRSF1A Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7132
Molecular Weight 21 kDa
Name TNF RI/TNFRSF1A
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
636R1025CF R&D Systems
RDS636R1025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
636R1025 R&D Systems
RDS636R1025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94
Specifications

Description

Source: E. coli-derived, Ile22-Thr211, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7132
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7132
TNF RI/TNFRSF1A
25μg
Human