missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TNF RI/TNFRSF1A Fc Chimera Recombinant Protein

$404.12 - $404.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7132
Molecular Weight 48 kDa (monomer)
Name TNF RI/TNFRSF1A Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
372RI050 R&D Systems
RDS372RI050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $404.12
372RI050CF R&D Systems
RDS372RI050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $404.12
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu30-Thr211 & Asp41-Thr211, both with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 48 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7132
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
48 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7132
TNF RI/TNFRSF1A Fc Chimera
50μg
Human