missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TNF-alpha Recombinant Protein

$129.05 - $2,613.64

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7124
Molecular Weight 17.5 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
210TA005CF R&D Systems
RDS210TA005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $129.05
210TA02MCF R&D Systems
RDS210TA02MCF
Lyophilized from a 0.2μm filtered solution in PBS. 2mg Each for $2,613.64
210TA020CF R&D Systems
RDS210TA020CF
Lyophilized from a 0.2μm filtered solution in PBS. 20μg Each for $272.44
210TA100 R&D Systems
RDS210TA100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 100μg Each for $819.94
210TA100CF R&D Systems
RDS210TA100CF
Lyophilized from a 0.2μm filtered solution in PBS. 100μg Each for $819.94
210TA005 R&D Systems
RDS210TA005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $129.05
210TA020 R&D Systems
RDS210TA020
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 20μg Each for $272.44
Specifications

Description

Source: E. coli-derived, Val77-Leu233, with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Entrez Gene ID: 7124
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
7124
TNF-alpha
Yes