missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TLR4/MD-2 Complex Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7099
Molecular Weight 70.6 kDa (TLR4), 19.2 kDa (MD-2)
Name TLR4/MD-2 Complex
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3146TM050CF R&D Systems
RDS3146TM050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
3146TM050 R&D Systems
RDS3146TM050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Glu17-Asn160, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 70.6 kDa (TLR4), 19.2 kDa (MD-2)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7099
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
70.6 kDa (TLR4), 19.2 kDa (MD-2)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7099
TLR4/MD-2 Complex
50μg
Human