Please login to your online account to display your discounted pricing

R&D Systems Human TL1A/TNFSF15 Recombinant Protein


Manufacturer: r&d systems  RDS1319TL010

 View more versions of this product

Catalog No. 1319TL010

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 21 kDa (monomer)
Name TL1A/TNFSF15
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 9966