missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TGF-beta RII Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7048
Molecular Weight 15 kDa
Name TGF-beta RII
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
241R2025 R&D Systems
RDS241R2025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94
241R2025CF R&D Systems
RDS241R2025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ile24-Asp159
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7048
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7048
TGF-beta RII
25μg
Human