Please login to your online account to display your discounted pricing

R&D Systems Human TGF-beta 1 Recombinant Protein, (Human Cell-expressed)

$337.62 - $1,054.58

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7040
Molecular Weight 12.8 kDa (monomer)
Name TGF-beta 1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

7754BH005

 
r&d systems
RDS7754BH005
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 5μg Each for $337.62

7754BH005CF

 
r&d systems
RDS7754BH005CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 5μg Each for $337.62

7754BH025

 
r&d systems
RDS7754BH025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 25μg Each for $1,054.58

7754BH025CF

 
r&d systems
RDS7754BH025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 25μg Each for $1,054.58
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7040
Molecular Weight 12.8 kDa (monomer)
Name TGF-beta 1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Human embryonic kidney cell, HEK293-derived, Ala279-Ser390
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 7040
Regulatory Status: RUO