missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human TGF-beta 1 Protein

$376.73 - $2,253.86

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7040
Name TGF-beta 1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
100B001 R&D Systems
RDS100B001
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 1μg Each for $376.73
100B001CF R&D Systems
RDS100B001CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 1μg Each for $376.73
100B010 R&D Systems
RDS100B010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $2,253.86
100B010CF R&D Systems
RDS100B010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $2,253.86
Specifications

Description

Source: Human platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 7040
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
TGF-beta 1
Human
7040
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.