Please login to your online account to display your discounted pricing

R&D Systems Human TGF-beta 1.2 Recombinant Protein

$483.61 - $483.61

Specifications

Conjugate Unlabeled
Molecular Weight 12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
Name TGF-beta 1.2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 1μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

304B3001

 
r&d systems
RDS304B3001
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $483.61

304B3001CF

 
r&d systems
RDS304B3001CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $483.61
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
Name TGF-beta 1.2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 1μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala379-Ser390 (TGF-beta1) & Ala303-Ser414 (TGF-beta2)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO