Please login to your online account to display your discounted pricing

R&D Systems Human Sonic Hedgehog/Shh N-Terminus Recombinant Protein


Manufacturer: r&d systems  RDS1314SH025CF

 View more versions of this product

Catalog No. 1314SH025CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 20 kDa
Name Sonic Hedgehog/Shh N-Terminus
Purity 97%, by SDS-PAGE with silver staining, under reducing conditions.
Quantity 25μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation Supplied as a 0.2μm filtered solution in NaH2PO4, NaCl and DTT .
Gene ID (Entrez) 6469