missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human sFRP-1 (Histidine-tagged) Recombinant Protein

$423.65 - $423.65

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6422
Name sFRP-1 (Histidine-tagged)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
1384SF025CF R&D Systems
RDS1384SF025CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. 33.4 kDa & 32.5 kDa & 32.2 kDa Each for $423.65
1384SF025 R&D Systems
RDS1384SF025
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. 33 kDa Each for $423.65
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser32-Lys314 with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 33.4 kDa & 32.5 kDa & 32.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 6422
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
sFRP-1 (Histidine-tagged)
25μg
Human
6422
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes