missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human S5a/Angiocidin Biotin Recombinant Protein

$319.37 - $350.65

Specifications

Conjugate Biotin
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 5710
Name S5a/Angiocidin Biotin
Quantity 250μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Purity Price Quantity  
Catalog Number Mfr. No. Purity Price Quantity  
SP405250 R&D Systems
RDSSP405250
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. Each for $350.65
UBE305250 R&D Systems
RDSUBE305250
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $319.37
Specifications

Description

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 5710
Regulatory Status: RUO

Description & Specifications

Specifications

Biotin
5710
250μg
Human
Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
S5a/Angiocidin Biotin
Yes