Please login to your online account to display your discounted pricing

R&D Systems Human S5a/Angiocidin Biotin Recombinant Protein


Manufacturer: r&d systems  RDSUBE305250

 View more versions of this product

Catalog No. UBE305250

  • / Each
 

Description & Specifications

Specifications

Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 5710
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Human
Recombinant Yes
Conjugate Biotin
Name S5a/Angiocidin Biotin
Quantity 250μg