Please login to your online account to display your discounted pricing

R&D Systems Human Ret Recombinant Protein


Manufacturer: r&d systems  RDS1168CR050

 View more versions of this product

Catalog No. 1168CR050

  • / Each
 

Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Name Ret
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Human
Recombinant Yes
Quantity 50μg
Molecular Weight 69 kDa
Conjugate Unlabeled
Gene ID (Entrez) 5979