Please login to your online account to display your discounted pricing

R&D Systems Human Ret Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5979
Molecular Weight 69 kDa
Name Ret
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1168CR050CF

 
r&d systems
RDS1168CR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81

1168CR050

 
r&d systems
RDS1168CR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5979
Molecular Weight 69 kDa
Name Ret
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Leu29-Arg635, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 69 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5979
Regulatory Status: RUO