Please login to your online account to display your discounted pricing

R&D Systems Human Ret Recombinant Protein


Manufacturer: r&d systems  RDS1168CR050CF

 View more versions of this product

Catalog No. 1168CR050CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 69 kDa
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Gene ID (Entrez) 5979
Quantity 50μg
Source Human
Recombinant Yes
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Name Ret