missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Relaxin-3 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 117579
Molecular Weight 2.46 kDa (A chain), 3.04 kDa (B chain)
Name Relaxin-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3107RN025 R&D Systems
RDS3107RN025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
3107RN025CF R&D Systems
RDS3107RN025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Specifications

Description

Source: E. coli-derived, Asp119-Cys142 (A chain) & Ala27-Arg53 (B chain)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 2.46 kDa (A chain), 3.04 kDa (B chain)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 117579
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
2.46 kDa (A chain), 3.04 kDa (B chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
117579
Relaxin-3
25μg
Human