missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Relaxin-2 (B-33/A-24) Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6019
Molecular Weight 2.7 kDa (A chain), 3.9 kDa (B chain)
Name Relaxin-2 (B-33/A-24)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3596RN025CF R&D Systems
RDS3596RN025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $415.83
3596RN025 R&D Systems
RDS3596RN025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $415.83
Specifications

Description

Source: E. coli-derived, Val23-Arg55 (B chain), with an N-terminal Met & Gln162-Cys185 (A chain)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 2.7 kDa (A chain), 3.9 kDa (B chain)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6019
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
2.7 kDa (A chain), 3.9 kDa (B chain)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6019
Relaxin-2 (B-33/A-24)
25μg
Human