Please login to your online account to display your discounted pricing

R&D Systems Human Relaxin-2 (B-29/A-24) Recombinant Protein

$407.05 - $407.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6019
Molecular Weight 2.7 kDa (A chain); 3.3 kDa (B chain)
Name Relaxin-2 (B-29/A-24)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

6586RN025CF

 
r&d systems
RDS6586RN025CF
Lyophilized from a 0.2μm filtered solution in Sodium Acetate. Each for $407.05

6586RN025

 
r&d systems
RDS6586RN025
Lyophilized from a 0.2μm filtered solution in sodium acetate with BSA as a carrier protein. Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6019
Molecular Weight 2.7 kDa (A chain); 3.3 kDa (B chain)
Name Relaxin-2 (B-29/A-24)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Asp25-Ser53 (B chain) & Gln162-Cys185 (A chain)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 2.7 kDa (A chain); 3.3 kDa (B chain)
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 6019
Regulatory Status: RUO