missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Relaxin-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6013
Molecular Weight 2.8 kDa (A chain), 3.3 kDa (B chain)
Name Relaxin-1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3257RN025CF R&D Systems
RDS3257RN025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
3257RN025 R&D Systems
RDS3257RN025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Arg162-Cys185 (A chain) & Lys26-Ser53 (B chain), with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 2.8 kDa (A chain), 3.3 kDa (B chain)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6013
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
2.8 kDa (A chain), 3.3 kDa (B chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6013
Relaxin-1
25μg
Human