Please login to your online account to display your discounted pricing

R&D Systems Human R-Spondin 4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 343637
Molecular Weight 24.1 kDa (monomer)
Name R-Spondin 4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

4575RS025

 
r&d systems
RDS4575RS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

4575RS025CF

 
r&d systems
RDS4575RS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 343637
Molecular Weight 24.1 kDa (monomer)
Name R-Spondin 4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-Pro234
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 343637
Regulatory Status: RUO