missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human R-Spondin 2 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 340419
Molecular Weight 22 kDa
Name R-Spondin 2
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3266RS025CF R&D Systems
RDS3266RS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
3266RS025 R&D Systems
RDS3266RS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln22-Gly205, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 340419
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
340419
R-Spondin 2
25μg
Human