Please login to your online account to display your discounted pricing

R&D Systems Human Protein Fraction II Protein Fraction


Manufacturer: r&d systems  RDSF37005M

 View more versions of this product

Catalog No. F37005M

  • / Each
 

Description & Specifications

Specifications

Name Protein Fraction II
Formulation Supplied as a solution in HEPES.
Conjugate Unlabeled
Source Human
Quantity 5mg