Please login to your online account to display your discounted pricing

R&D Systems Human Protein Fraction I Protein Fraction


Manufacturer: r&d systems  RDSF37501M

 View more versions of this product

Catalog No. F37501M

  • / Each
 

Description & Specifications

Specifications

Name Protein Fraction I
Source Human
Conjugate Unlabeled
Quantity 1mg
Formulation Supplied as a solution in HEPES.