Please login to your online account to display your discounted pricing

R&D Systems Human Protein Fraction I Protein Fraction

$142.08 - $559.23

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES.
Name Protein Fraction I
Source Human
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

F37501M

 
r&d systems
RDSF37501M
1mg Each for $142.08

F37505M

 
r&d systems
RDSF37505M
5mg Each for $559.23
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES.
Name Protein Fraction I
Source Human

Source: Human cervical epithelial carcinoma cell line, HeLa-derived
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO