missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Pro EGF aa 21-1023 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1950
Molecular Weight 112 kDa
Name Pro EGF aa 21-1023
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4289EG025 R&D Systems
RDS4289EG025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
4289EG025CF R&D Systems
RDS4289EG025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Met1-Arg1023, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 112 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1950
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
112 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
1950
Pro EGF aa 21-1023
25μg
Human