missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human PlGF Recombinant Protein

$389.76 - $1,192.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5228
Molecular Weight 14.5 kDa (monomer)
Name PlGF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
264PG010CF R&D Systems
RDS264PG010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $389.76
264PG050 R&D Systems
RDS264PG050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $1,192.76
264PG010 R&D Systems
RDS264PG010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $389.76
264PG050CF R&D Systems
RDS264PG050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $1,192.76
Specifications

Description

Source: E. coli-derived, Ala21-Arg149
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 5228
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14.5 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
5228
PlGF
Yes