missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Pleiotrophin/PTN Recombinant Protein

$376.73 - $1,080.65

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 5764
Molecular Weight 15.3 kDa
Name Pleiotrophin/PTN
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
252PL250 R&D Systems
RDS252PL250
250μg Each for $1,080.65
252PL050 R&D Systems
RDS252PL050
50μg Each for $376.73
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly33-Asp168
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5764
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
5764
Pleiotrophin/PTN
Yes
Lyophilized from a 0.2μm filtered solution in PBS.
15.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human