missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human PGRP-S Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8993
Molecular Weight 20.2 kDa
Name PGRP-S
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2590PG050 R&D Systems
RDS2590PG050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
2590PG050CF R&D Systems
RDS2590PG050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln22-Pro196, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8993
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8993
PGRP-S
50μg
Human