missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Pentraxin 3/TSG-14 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5806
Molecular Weight 41 kDa (monomer)
Name Pentraxin 3/TSG-14
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1826TS025 R&D Systems
RDS1826TS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1826TS025CF R&D Systems
RDS1826TS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Glu18-Ser381, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 41 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5806
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
41 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
5806
Pentraxin 3/TSG-14
25μg
Human