Please login to your online account to display your discounted pricing

R&D Systems Human Pentraxin 2/SAP Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 325
Molecular Weight 25 kDa (monomer)
Name Pentraxin 2/SAP
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1948SA050

 
r&d systems
RDS1948SA050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

1948SA050CF

 
r&d systems
RDS1948SA050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 325
Molecular Weight 25 kDa (monomer)
Name Pentraxin 2/SAP
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, His20-Val223, with a C-terminal 10-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 325
Regulatory Status: RUO