missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human PDGF R alpha Recombinant Protein

$467.98 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5156
Molecular Weight 56 kDa
Name PDGF R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
322PR050 R&D Systems
RDS322PR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.98
322PR050CF R&D Systems
RDS322PR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.98
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln24-Glu524
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 56 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5156
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
56 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
5156
PDGF R alpha
50μg
Human