Please login to your online account to display your discounted pricing

R&D Systems Human PDGF Protein


Manufacturer: r&d systems  RDS120HD001

 View more versions of this product

Catalog No. 120HD001

  • / Each
 

Description & Specifications

Specifications

Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 5154
Structural Form Mostly disulfide-linked heterodimer
Name PDGF
Conjugate Unlabeled
Quantity 1μg
Source Human