Please login to your online account to display your discounted pricing

R&D Systems Human PDGF Protein

$381.54 - $1,376.82

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 5154
Name PDGF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

120HD005

 
r&d systems
RDS120HD005
5μg Each for $1,376.82

120HD001

 
r&d systems
RDS120HD001
1μg Each for $381.54
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 5154
Name PDGF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Human

Source: Human platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 5154
Regulatory Status: RUO