Please login to your online account to display your discounted pricing

R&D Systems Human PDGF Protein


Manufacturer: r&d systems  RDS120HD005

 View more versions of this product

Catalog No. 120HD005

  • / Each
 

Description & Specifications

Specifications

Name PDGF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 5μg
Source Human
Structural Form Mostly disulfide-linked heterodimer
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 5154