missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human PDGF-DD Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 80310
Molecular Weight 14 kDa (monomer)
Name PDGF-DD
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1159SB025CF R&D Systems
RDS1159SB025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $416.79
1159SB025 R&D Systems
RDS1159SB025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser250-Arg370
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 80310
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
80310
PDGF-DD
25μg
Human