missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human PDGF-CC Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56034
Molecular Weight 13.4 kDa (monomer)
Name PDGF-CC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1687CC025 R&D Systems
RDS1687CC025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $415.83
1687CC025CF R&D Systems
RDS1687CC025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $415.83
Specifications

Description

Source: E. coli-derived, Val235-Gly345, with an N-terminal Met and 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.4 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 56034
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
13.4 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
56034
PDGF-CC
25μg
Human