missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human PD-ECGF/Thymidine Phosphorylase Recombinant Protein

$311.55 - $1,067.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1890
Molecular Weight 49 kDa
Name PD-ECGF/Thymidine Phosphorylase
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
229PE010 R&D Systems
RDS229PE010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $311.55
229PE010CF R&D Systems
RDS229PE010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $311.55
229PE050 R&D Systems
RDS229PE050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $1,067.62
229PE050CF R&D Systems
RDS229PE050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $1,067.62
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala11-Gln482
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 49 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1890
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
49 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
1890
PD-ECGF/Thymidine Phosphorylase
Yes