missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human P-Selectin/CD62P Recombinant Protein

$324.58 - $885.12

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Gene ID (Entrez) 6403
Molecular Weight 80 kDa
Name P-Selectin/CD62P
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ADP3200 R&D Systems
RDSADP3200
200μg Each for $885.12
ADP3050 R&D Systems
RDSADP3050
50μg Each for $324.58
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Trp42-Ala771
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 80 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Entrez Gene ID: 6403
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
6403
P-Selectin/CD62P
Yes
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
80 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human