missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Osteoprotegerin/TNFRSF11B Recombinant Protein

$467.46 - $467.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4982
Molecular Weight 43.5 kDa (monomer)
Name Osteoprotegerin/TNFRSF11B
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
185OS025CF R&D Systems
RDS185OS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.46
185OS025 R&D Systems
RDS185OS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Glu22-Leu401
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 43.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4982
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
43.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
4982
Osteoprotegerin/TNFRSF11B
25μg
Human