missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human NT-3 Recombinant Protein

$298.51 - $866.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4908
Molecular Weight 13.7 kDa
Name NT-3
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
267N3005 R&D Systems
RDS267N3005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $298.51
267N3005CF R&D Systems
RDS267N3005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $298.51
267N3025 R&D Systems
RDS267N3025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $866.87
267N3025CF R&D Systems
RDS267N3025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $866.87
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Tyr139-Thr257 (K196R)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4908
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
13.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
4908
NT-3
Yes