Please login to your online account to display your discounted pricing

R&D Systems Human NGF R/TNFRSF16 Fc Chimera Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4804
Molecular Weight 51 kDa (monomer)
Name NGF R/TNFRSF16 Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

367NR050CF

 
r&d systems
RDS367NR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81

367NR050

 
r&d systems
RDS367NR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4804
Molecular Weight 51 kDa (monomer)
Name NGF R/TNFRSF16 Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Lys29-Asn250
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 51 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4804
Regulatory Status: RUO